Thermo Scientific Chemicals

(3-Cyanopropyl)triethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: L05367.22
100 g, Each
Thermo Scientific Chemicals

(3-Cyanopropyl)triethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: L05367.22
100 g, Each
Quantity
Catalog number: L05367.22
also known as L05367-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1067-47-6
IUPAC Name
4-(triethoxysilyl)butanenitrile
Molecular Formula
C10H21NO3Si
InChI Key
VGIURMCNTDVGJM-UHFFFAOYSA-N
SMILES
CCO[Si](CCCC#N)(OCC)OCC
Assay (GC)
≥96.0%
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4145-1.4195 @ 20?C

Description

(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.

Solubility
Not miscible in water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text