Thermo Scientific Chemicals

1-Bromo-2-(2-methoxyethoxy)ethane, tech. 90%, stab. with sodium carbonate, Thermo Scientific Chemicals

Catalog number: L05406.06
5 g, Each
Thermo Scientific Chemicals

1-Bromo-2-(2-methoxyethoxy)ethane, tech. 90%, stab. with sodium carbonate, Thermo Scientific Chemicals

Catalog number: L05406.06
5 g, Each
Quantity
Catalog number: L05406.06
also known as L05406-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54149-17-6
IUPAC Name
1-bromo-2-(2-methoxyethoxy)ethane
Molecular Formula
C5H11BrO2
InChI Key
HUXJXNSHCKHFIL-UHFFFAOYSA-N
SMILES
COCCOCCBr
Form
Liquid
Refractive Index
1.4540-1.4600 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
>88.0%

Description

1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store at 4°C. Protect from heat. Store away from strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text