Thermo Scientific Chemicals

3-Octyn-1-ol, 96%, Thermo Scientific Chemicals

Catalog number: L05414.06
5 g, Each
Thermo Scientific Chemicals

3-Octyn-1-ol, 96%, Thermo Scientific Chemicals

Catalog number: L05414.06
5 g, Each
Quantity
Catalog number: L05414.06
also known as L05414-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14916-80-4
IUPAC Name
oct-3-yn-1-ol
Molecular Formula
C8H14O
InChI Key
LRZGRGVRZSDRTK-UHFFFAOYSA-N
SMILES
CCCCC#CCCO
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4535-1.4600 @ 20?C
Form
Liquid
Assay (GC)
≥95.0%

Description

3-Octyn-1-ol, is used for a study of the reduction of acetylenic compounds, including this alcohol, to (E)-alkenes by alkali metals in liquid ammonia. Some other applications includes as corrosion inhibitor formulation, metal complex solution, solvent stabilizer, biodegradable chemical intermediate to make pharmaceuticals, chemical compound synthesis and reagents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octyn-1-ol, is used for a study of the reduction of acetylenic compounds, including this alcohol, to (E)-alkenes by alkali metals in liquid ammonia. Some other applications includes as corrosion inhibitor formulation, metal complex solution, solvent stabilizer, biodegradable chemical intermediate to make pharmaceuticals, chemical compound synthesis and reagents

Solubility
It is soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text