4-Ethoxybenzhydrazide, 98+%, Thermo Scientific Chemicals
4-Ethoxybenzhydrazide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethoxybenzhydrazide, 98+%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 g
Catalog number L05433.06
also known as L05433-06
Price (USD)/ Each
76.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
76.00
Add to cart
4-Ethoxybenzhydrazide, 98+%, Thermo Scientific Chemicals
Catalog numberL05433.06
Price (USD)/ Each
76.00
-
Add to cart
Chemical Identifiers
CAS58586-81-5
IUPAC Name4-ethoxybenzohydrazide
Molecular FormulaC9H12N2O2
InChI KeySTTILZBQLOSDNQ-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)C(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥98.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only