Thermo Scientific Chemicals

2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals

Catalog number: L05442.14
25 g, Each
Thermo Scientific Chemicals

2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals

Catalog number: L05442.14
25 g, Each
Quantity
Catalog number: L05442.14
also known as L05442-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5271-38-5
IUPAC Name
2-(methylsulfanyl)ethan-1-ol
Molecular Formula
C3H8OS
InChI Key
WBBPRCNXBQTYLF-UHFFFAOYSA-N
SMILES
CSCCO
Form
Liquid
Refractive Index
1.4930-1.4960 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
>98.5%

Description

2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Not miscible in water.

Notes
Do not store together with acids. Store away from Oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text