1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC. It may be employed as reagent for the fluorometric determination of airborne diisocyantes. It is a compound that belongs to a class of selective α2-adrenoceptor antagonists. 1-(2-Pyridyl)piperazine shows sympatholytic activity. 1-(2-Pyridyl)piperazine is also a metabolite of Azaperone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(2-Pyridyl)piperazine may be employed as reagent for the determination of both aliphatic and aromatic isocyanates in air by reversed-phase HPLC. It may be employed as reagent for the fluorometric determination of airborne diisocyantes. It is a compound that belongs to a class of selective α2-adrenoceptor antagonists. 1-(2-Pyridyl)piperazine shows sympatholytic activity. 1-(2-Pyridyl)piperazine is also a metabolite of Azaperone.
Solubility
Soluble in Chloroform, Methanol.
Notes
Air Sensitive. Store away from air and oxidizing agents. Incompatible with carbondioxide, Strong acids, Acid chlorides, Acid anhydrides.
RUO – Research Use Only
P.A. Goldberg; R.F. Walker; P.A. Ellwood; H.L. Hardy. Determination of trace atmospheric isocyanate concentrations by reversed-phase high-performance liquid chromatography using 1-(2-pyridyl)piperazine reagent.Journal of Chromatography A.1981,212 93-104.
T. Salthammer; C. Wismach; H. Miertzsch. Absorption and fluorescence of 1-(2-pyridyl)-piperazine and four diisocyanate derivatives in solution.Journal of Photochemistry and Photobiology A: Chemistry.1997,107 159-164.