Molecular sieves, 4A, -8+12 (ca 2mm) beads, Thermo Scientific Chemicals
Molecular sieves, 4A, -8+12 (ca 2mm) beads, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves, 4A, -8+12 (ca 2mm) beads, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
250 g
1000 g
Catalog number L05454.0I
also known as L05454-0I
Price (USD)/ Each
305.65
Online exclusive
339.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
305.65
Online exclusive
339.00 
Save 33.35 (10%)
Add to cart
Molecular sieves, 4A, -8+12 (ca 2mm) beads, Thermo Scientific Chemicals
Catalog numberL05454.0I
Price (USD)/ Each
305.65
Online exclusive
339.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS70955-01-0
IUPAC NameMolecular sieves
Synonym2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
CommentBulk Density ≥680 g/L
CommentWater absorption (50% rH, 20?C, 24hrs) ≥20%
FormBeads
CommentResidual water (550?C, 2hrs) ≤1.5%
Appearance (Color)Pale grey to brown
View more
Molecular sieves, 4 Å is used to absorb molecules as large as butane. It is utilized for the separation of natural gas, liquified petroleum gas (LPG) and alkenes. It finds application in the refining and purification of liquid and gases. It is also used for drying and purification of air, inert gases and solvents. It is actively involved to remove methanol, carbon dioxide and ammonia from fluid streams. It plays a vital role to remove heavy metal ions available in water. It is also used as a soap forming agent and as a detergent auxiliary. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only