Thermo Scientific Chemicals

4-Chloro-1,8-naphthalic anhydride, 94%, Thermo Scientific Chemicals

Catalog number: L05508.36
500 g, Each
Thermo Scientific Chemicals

4-Chloro-1,8-naphthalic anhydride, 94%, Thermo Scientific Chemicals

Catalog number: L05508.36
500 g, Each
Quantity
Catalog number: L05508.36
also known as L05508-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4053-08-1
IUPAC Name
10-chloro-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Formula
C12H5ClO3
InChI Key
UJEUBSWHCGDJQU-UHFFFAOYSA-N
SMILES
ClC1=C2C=CC=C3C(=O)OC(=O)C(C=C1)=C23
Form
Powder or lumps
Comment
Material sourced in UK and US
Assay from Supplier's CofA
≥92.0% (US-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)
Appearance (Color)
Pale cream to cream to pale brown

Description

4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store at 4°C. Store away from moisture. Incompatible with oxidizing agents, water, Strong bases, Strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text