Thermo Scientific Chemicals

Molecular sieves, 4A, 0.4-0.8mm (0.02-0.03in) beads, Thermo Scientific Chemicals

Catalog number: L05512.30
250 g, Each
Thermo Scientific Chemicals

Molecular sieves, 4A, 0.4-0.8mm (0.02-0.03in) beads, Thermo Scientific Chemicals

Catalog number: L05512.30
250 g, Each
Quantity
Catalog number: L05512.30
also known as L05512-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
70955-01-0
IUPAC Name
Molecular sieves
Synonym
2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL Number
MFCD00131613
Appearance (Color)
Pale brown
Form
beads
Comment
Absorption capacity ca. 20%
Particle Size
0.4-0.8 mm

Description

Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads are useful in general gas drying applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text