Thermo Scientific Chemicals

3-Butyn-2-one, 98%, Thermo Scientific Chemicals

Catalog number: L05527.03
1 g, Each
Thermo Scientific Chemicals

3-Butyn-2-one, 98%, Thermo Scientific Chemicals

Catalog number: L05527.03
1 g, Each
Quantity
Catalog number: L05527.03
also known as L05527-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1423-60-5
Assay (GC)
≥97.5%
Refractive Index
1.4045-1.4085 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Identification (FTIR)
Conforms

Description

3-Butyn-2-one was used in the synthesis of clerodane diterpenoid (±)-sacacarin. It was used as substrate in stereoselective, conjugate arylation mediated by gallium(III) chloride leading to (E)-α,β-unsaturated ketones. 3-Butyn-2-one undergoes asymmetric double-Michael reaction with ortho-tosylamidophenyl malonate catalyzed by chiral aminophosphines to yield indolines2. It undergoes double Michael reaction with nitrogen-containing tethered diacid to give pipecolic acid derivatives. 3-Butyn-2-one, is used as a reactant with Dimethyl­ acetone-1,3-dicarboxyl­ate, under mild conditions to give a benzenoid product via a Michael addition - aldol cyclization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Butyn-2-one was used in the synthesis of clerodane diterpenoid (+/-)-sacacarin. It was used as substrate in stereoselective, conjugate arylation mediated by gallium(III) chloride leading to (E)-α,β-unsaturated ketones. 3-Butyn-2-one undergoes asymmetric double-Michael reaction with ortho-tosylamidophenyl malonate catalyzed by chiral aminophosphines to yield indolines2. It undergoes double Michael reaction with nitrogen-containing tethered diacid to give pipecolic acid derivatives. 3-Butyn-2-one, is used as a reactant with Dimethyl­ acetone-1,3-dicarboxyl­ate, under mild conditions to give a benzenoid product via a Michael addition - aldol cyclization process.

Solubility
It is soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text