Thermo Scientific Chemicals

N,N-Dimethylbutyramide, 98%, Thermo Scientific Chemicals

Catalog number: L05556.06
5 g, Each
Thermo Scientific Chemicals

N,N-Dimethylbutyramide, 98%, Thermo Scientific Chemicals

Catalog number: L05556.06
5 g, Each
Quantity
Catalog number: L05556.06
also known as L05556-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
760-79-2
IUPAC Name
N,N-dimethylbutanamide
Molecular Formula
C6H13NO
InChI Key
VIJUZNJJLALGNJ-UHFFFAOYSA-N
SMILES
CCCC(=O)N(C)C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4405-1.4445 @ 20?C
Identification (FTIR)
Conforms

Description

N,N-Dimethylbutyramide, is used as an organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylbutyramide, is used as an organic intermediate, pharmaceutical intermediate.

Solubility
Very Soluble in water (110 g/L) (25°C).

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text