Thermo Scientific Chemicals

n-Pentyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L05563.06
5 g, Each
Thermo Scientific Chemicals

n-Pentyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L05563.06
5 g, Each
Quantity
Catalog number: L05563.06
also known as L05563-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6521-29-5
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or chunks or fused solid
Identification (FTIR)
Conforms

Description

n-Pentyl 4-hydroxybenzoate, is used as an antimicrobial agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Pentyl 4-hydroxybenzoate, is used as an antimicrobial agent in cosmetic products.

Solubility
Insoluble in water. Slightly soluble in chloroform, methanol.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text