Thermo Scientific Chemicals

Potassium tetrakis(4-chlorophenyl)borate, 98%, Thermo Scientific Chemicals

Catalog number: L05609.03
1 g, Each
Thermo Scientific Chemicals

Potassium tetrakis(4-chlorophenyl)borate, 98%, Thermo Scientific Chemicals

Catalog number: L05609.03
1 g, Each
Quantity
Catalog number: L05609.03
also known as L05609-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14680-77-4
IUPAC Name
boron(3+) potassium tetrakis(3-chlorobenzen-1-ide)
Molecular Formula
C24H16BCl4K
InChI Key
VHPYLDPFVZBNQT-UHFFFAOYSA-N
SMILES
[B+3].[K+].ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1.ClC1=CC=C[C-]=C1
Appearance (Color)
White
Form
Powder
Assay (HPLC)
≥97.5%
Melting Point
>300?C

Description

Potassium tetrakis(4-chlorophenyl)borate is used as lipophilic anionic site in constructing fluorescent optode for sodium. It was also used as coating for the piezoelectric crystal in quantifying Potassium using quartz crystal microbalance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrakis(4-chlorophenyl)borate is used as lipophilic anionic site in constructing fluorescent optode for sodium. It was also used as coating for the piezoelectric crystal in quantifying Potassium using quartz crystal microbalance.

Solubility
Soluble in chloroform.

Notes
Hygroscopic. Store under dry inert gas. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text