Thermo Scientific Chemicals

2-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: L05627.14
25 g, Each
Thermo Scientific Chemicals

2-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: L05627.14
25 g, Each
Quantity
Catalog number: L05627.14
also known as L05627-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2706-56-1
IUPAC Name
2-(pyridin-2-yl)ethan-1-amine
Molecular Formula
C7H10N2
InChI Key
XPQIPUZPSLAZDV-UHFFFAOYSA-N
SMILES
NCCC1=CC=CC=N1
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.5335-1.5385 @20°C (non-U.S. specification)
Appearance (Color)
Clear, colorless to yellow
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms (non-U.S. specification)

Description

2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.

Solubility
Soluble in water (partly), and most common polar solvent. Insoluble in non-polar solvents, and oils.

Notes
Air Sensitive. Store at 4°C. Stor in the dark. Incompatible with air, oxidizing agents, carbondioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text