Thermo Scientific Chemicals

Methyl 4-amino-3-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L05687.09
10 g, Each
Thermo Scientific Chemicals

Methyl 4-amino-3-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L05687.09
10 g, Each
Quantity
Catalog number: L05687.09
also known as L05687-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63435-16-5
IUPAC Name
methyl 3-amino-4-hydroxybenzoate
Molecular Formula
C8H9NO3
InChI Key
VNQABZCSYCTZMS-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(O)C(N)=C1
Assay (GC)
≥97.5%
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
117.0-123.0?C
Appearance (Color)
Pale yellow to yellow to brown
Form
Crystals or powder or crystalline powder

Description

Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Notes
Incompatible with Strong oxidizing agents, Strong acids and bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text