Thermo Scientific Chemicals

4-Chloro-1-methylpiperidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L05699.14
25 g, Each
Thermo Scientific Chemicals

4-Chloro-1-methylpiperidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L05699.14
25 g, Each
Quantity
Catalog number: L05699.14
also known as L05699-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5382-23-0
IUPAC Name
hydrogen 4-chloro-1-methylpiperidine chloride
Molecular Formula
C6H13Cl2N
InChI Key
QDOJNWFOCOWAPO-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN1CCC(Cl)CC1
Appearance (Color)
White to cream
Form
Powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Melting Point (clear melt)
152-166?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text