Thermo Scientific Chemicals

2-Amino-5-chlorothiazole hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L05707.03
1 g, Each
Thermo Scientific Chemicals

2-Amino-5-chlorothiazole hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L05707.03
1 g, Each
Quantity
Catalog number: L05707.03
also known as L05707-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55506-37-1
IUPAC Name
hydrogen 5-chloro-1,3-thiazol-2-amine chloride
Molecular Formula
C3H4Cl2N2S
InChI Key
GTMGFQYVLSQTKP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1=NC=C(Cl)S1
Form
Powder or granules
Appearance (Color)
Dark cream to cream to pale brown or pale pink to gray
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.

Solubility
Soluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text