trans-4-Decenal, 96%, may cont. ca 7% cis-isomer, Thermo Scientific Chemicals
trans-4-Decenal, 96%, may cont. ca 7% cis-isomer, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-Decenal, 96%, may cont. ca 7% cis-isomer, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05715.06
also known as L05715-06
Price (USD)/ Each
48.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
48.10
Add to cart
trans-4-Decenal, 96%, may cont. ca 7% cis-isomer, Thermo Scientific Chemicals
Catalog numberL05715.06
Price (USD)/ Each
48.10
-
Add to cart
Chemical Identifiers
CAS65405-70-1
IUPAC Name(4E)-dec-4-enal
Molecular FormulaC10H18O
InChI KeyCWRKZMLUDFBPAO-VOTSOKGWSA-N
SMILESCCCCCC=CCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Impurity contentcis-isomer typically 6.0 - 8.5%
Appearance (Color)Clear colourless to pale yellow
Assay (GC)> 95.0% (sum of isomers)
Free acid (titration)< 4.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only