Thermo Scientific Chemicals

4-(2-Chloroethyl)morpholine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L05771.22
100 g, Each
Thermo Scientific Chemicals

4-(2-Chloroethyl)morpholine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L05771.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L05771.22
also known as L05771-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3647-69-6
IUPAC Name
hydrogen 4-(2-chloroethyl)morpholine chloride
Molecular Formula
C6H13Cl2NO
InChI Key
NBJHDLKSWUDGJG-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCCN1CCOCC1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Solution Test
Clear (5% in water)

Description

4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.

Solubility
Soluble in water, and methanol.

Notes
Hygroscopic. Do not store together with acids. Store under dry inert gas. Protect from humidity and water. Incompatible materials are acids, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text