Thermo Scientific Chemicals

2-Amino-5-cyclopropyl-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals

Catalog number: L05778.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-5-cyclopropyl-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals

Catalog number: L05778.06
5 g, Each
Quantity
Catalog number: L05778.06
also known as L05778-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57235-50-4
Purity
≥97.5%
Appearance (Color)
Off-white to pale cream
Mass Spec
Conforms
Form
Powder or crystals or crystalline powder

Description

2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.

Solubility
Soluble in Methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text