Thermo Scientific Chemicals

2-Amino-4-imino-2-thiazoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L05844.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-4-imino-2-thiazoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: L05844.06
5 g, Each
Quantity
Catalog number: L05844.06
also known as L05844-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36518-76-0
IUPAC Name
2-imino-2,5-dihydro-1,3-thiazol-4-amine hydrochloride
Molecular Formula
C3H6ClN3S
InChI Key
ANDLBJXZBLTCDP-UHFFFAOYSA-N
SMILES
Cl.NC1=NC(=N)SC1
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Appearance (Color)
White to cream
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text