Thermo Scientific Chemicals

3-Chloropropionitrile, 98%, Thermo Scientific Chemicals

Catalog number: L05902.14
25 g, Each
Thermo Scientific Chemicals

3-Chloropropionitrile, 98%, Thermo Scientific Chemicals

Catalog number: L05902.14
25 g, Each
Quantity
Catalog number: L05902.14
also known as L05902-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
542-76-7
IUPAC Name
3-chloropropanenitrile
Molecular Formula
C3H4ClN
InChI Key
GNHMRTZZNHZDDM-UHFFFAOYSA-N
SMILES
ClCCC#N
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4370-1.4395 @ 20?C

Description

3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.

Solubility
Soluble in water. (4.5g/100mL, 25°C). Miscible in Ether, Alcohols, Benzene, Acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text