3-Chloropropionitrile, 98%, Thermo Scientific Chemicals
3-Chloropropionitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloropropionitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number L05902.22
also known as L05902-22
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
3-Chloropropionitrile, 98%, Thermo Scientific Chemicals
Catalog numberL05902.22
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS542-76-7
IUPAC Name3-chloropropanenitrile
Molecular FormulaC3H4ClN
InChI KeyGNHMRTZZNHZDDM-UHFFFAOYSA-N
SMILESClCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4370-1.4395 @ 20?C
FormLiquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.

Solubility
Soluble in water. (4.5g/100mL, 25°C). Miscible in Ether, Alcohols, Benzene, Acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.
RUO – Research Use Only

General References:

  1. S. Heininger. Notes - Reactions of 3-Chloropropionitrile and Alkyl 3-Chloropropionates with Amines.J. Org. Chem.1957,22 (6), 704-706.
  2. J.R. Durig; G.A. Guirgis; A.S. Drew. Conformational analysis, ab initio calculations, and vibrational assignment of 3-chloropropionitrile.Journal of Molecular Structure.1994,327 55-69.