Thermo Scientific Chemicals

4,4'-Dimethylbenzhydrol, 98%, Thermo Scientific Chemicals

Catalog number: L05904.06
5 g, Each
Thermo Scientific Chemicals

4,4'-Dimethylbenzhydrol, 98%, Thermo Scientific Chemicals

Catalog number: L05904.06
5 g, Each
Quantity
Catalog number: L05904.06
also known as L05904-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
885-77-8
IUPAC Name
bis(4-methylphenyl)methanol
Molecular Formula
C15H16O
InChI Key
RGYZQSCFKFDECZ-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C(O)C1=CC=C(C)C=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
68.0-75.0?C
Appearance (Color)
White

Description

4,4'-Dimethylbenzhydrol is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethylbenzhydrol is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text