Thermo Scientific Chemicals

1-Methylpyrrole-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: L05906.03
1 g, Each
Thermo Scientific Chemicals

1-Methylpyrrole-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: L05906.03
1 g, Each
Quantity
Catalog number: L05906.03
also known as L05906-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6973-60-0
IUPAC Name
1-methyl-1H-pyrrole-2-carboxylic acid
Molecular Formula
C6H7NO2
InChI Key
ILAOVOOZLVGAJF-UHFFFAOYSA-N
SMILES
CN1C=CC=C1C(O)=O
Appearance (Color)
White to cream to yellow to brown to gray
Form
Crystals or powder or crystalline powder or lumps
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text