Thermo Scientific Chemicals

Ethylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: L05908.0F
2500 mL, Each
Thermo Scientific Chemicals

Ethylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: L05908.0F
2500 mL, Each
Quantity
Catalog number: L05908.0F
also known as L05908-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-41-4
IUPAC Name
ethylbenzene
Molecular Formula
C8H10
InChI Key
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
SMILES
CCC1=CC=CC=C1
Form
Liquid
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless
Refractive Index
1.4940-1.4970 @ 20°C (non-U.S. specification)

Description

Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.

Solubility
Miscible with alcohol, ethylalcohol, benzene, ethyl ether, carbon tetrachloride, organic solvents, sulfur dioxide and ether. Slightly miscible with chloroform. Immiscible with water and ammonia.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text