Thermo Scientific Chemicals

Methyl 3,4,5-trihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L05926.18
50 g, Each
Thermo Scientific Chemicals

Methyl 3,4,5-trihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L05926.18
50 g, Each
Quantity
Catalog number: L05926.18
also known as L05926-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-24-1
IUPAC Name
methyl 3,4,5-trihydroxybenzoate
Molecular Formula
C8H8O5
InChI Key
FBSFWRHWHYMIOG-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(O)=C(O)C(O)=C1
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White to cream or pale pink
Form
Crystals or powder or crystalline powder

Description

Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text