Appearance (Color)White to cream to yellow to brown or grey
FormPowder or lumps
Assay (HPLC)≥97.5%
Water Content (Karl Fischer Titration)≤1.0%
Melting Point (clear melt)125-135°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.