Thermo Scientific Chemicals

L-Arginine methyl ester dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L05959.06
5 g, Each
Thermo Scientific Chemicals

L-Arginine methyl ester dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L05959.06
5 g, Each
Quantity
Catalog number: L05959.06
also known as L05959-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26340-89-6
IUPAC Name
dihydrogen methyl (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoate dichloride
Molecular Formula
C7H18Cl2N4O2
InChI Key
XQYZOBNLCUAXLF-XRIGFGBMSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Optical Rotation
+21.0 ? 1.0? (c=2.5 in methanol)
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text