Thermo Scientific Chemicals

Triethyl orthoacetate, 97%, Thermo Scientific Chemicals

Catalog number: L06004.AE
100 mL, Each
Thermo Scientific Chemicals

Triethyl orthoacetate, 97%, Thermo Scientific Chemicals

Catalog number: L06004.AE
100 mL, Each
Quantity
Catalog number: L06004.AE
also known as L06004-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-39-7
IUPAC Name
1,1,1-triethoxyethane
Molecular Formula
C8H18O3
InChI Key
NDQXKKFRNOPRDW-UHFFFAOYSA-N
SMILES
CCOC(C)(OCC)OCC
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.3950-1.3990 @ 20?C

Description

Triethyl orthoacetate is used as pharmaceutical and water scavenger. It is also used in organic synthesis to introduce the acetate group into an alcohol. It is involved in the Johnson-Claisen rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl orthoacetate is used as pharmaceutical and water scavenger. It is also used in organic synthesis to introduce the acetate group into an alcohol. It is involved in the Johnson-Claisen rearrangement.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text