5-Methyl-1-hexyne, 98%, Thermo Scientific Chemicals
5-Methyl-1-hexyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methyl-1-hexyne, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06014.06
also known as L06014-06
Price (USD)/ Each
59.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
59.10
Add to cart
5-Methyl-1-hexyne, 98%, Thermo Scientific Chemicals
Catalog numberL06014.06
Price (USD)/ Each
59.10
-
Add to cart
Chemical Identifiers
CAS2203-80-7
IUPAC Name5-methylhex-1-yne
Molecular FormulaC7H12
InChI KeyHKNANEMUCJGPMS-UHFFFAOYSA-N
SMILESCC(C)CCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.0%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms
View more
5-Methyl-1-hexyne is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1-hexyne is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

General References:

  1. Akiya Ogawa; Takuma Ikeda; Kouichi Kimura and Toshikazu Hirao. Highly Regio- and Stereocontrolled Synthesis of Vinyl Sulfides via Transition-Metal-Catalyzed Hydrothiolation of Alkynes with Thiols.J. Am. Chem. Soc.1999,121 (22), 5108-5114.
  2. Shin-ichi Kawaguchi; Shoko Nagata; Akihiro Nomoto; Motohiro Sonoda and Akiya Ogawa. A Highly Regioselective Palladium-Catalyzed Hydrophosphination of Alkynes Using a Diphosphine-Hydrosilane Binary System.J. Org. Chem.2008,73 (20), 7928-7933.