Thermo Scientific Chemicals

4-Chloro-alpha-methylphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L06020.14
25 g, Each
Thermo Scientific Chemicals

4-Chloro-alpha-methylphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L06020.14
25 g, Each
Quantity
Catalog number: L06020.14
also known as L06020-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
938-95-4
IUPAC Name
2-(4-chlorophenyl)propanoic acid
Molecular Formula
C9H9ClO2
InChI Key
YOZILQVNIWNPFP-UHFFFAOYNA-N
SMILES
CC(C(O)=O)C1=CC=C(Cl)C=C1
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
53.5-59.5?C
Appearance (Color)
White to cream

Description

4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text