4-Dimethylaminoazobenzene-4'-sulfonyl chloride, 98+%, Thermo Scientific Chemicals
4-Dimethylaminoazobenzene-4'-sulfonyl chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Dimethylaminoazobenzene-4'-sulfonyl chloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 mg
Catalog number L06057.ME
also known as L06057-ME
Price (USD)/ Each
86.65
Online exclusive
96.20 
Save 9.55 (10%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
86.65
Online exclusive
96.20 
Save 9.55 (10%)
Add to cart
4-Dimethylaminoazobenzene-4'-sulfonyl chloride, 98+%, Thermo Scientific Chemicals
Catalog numberL06057.ME
Price (USD)/ Each
86.65
Online exclusive
96.20 
Save 9.55 (10%)
-
Add to cart
Chemical Identifiers
CAS56512-49-3
IUPAC Name4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonyl chloride
Molecular FormulaC14H14ClN3O2S
InChI KeyVTVWTPGLLAELLI-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red
Assay (Titration ex Chloride)≥98.0% to ≤102.0%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only