Thermo Scientific Chemicals

4-Methoxyphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L06076.14
25 g, Each
Thermo Scientific Chemicals

4-Methoxyphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L06076.14
25 g, Each
Quantity
Catalog number: L06076.14
also known as L06076-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19501-58-7
IUPAC Name
(4-methoxyphenyl)hydrazine hydrochloridyl
Molecular Formula
C7H10ClN2O
InChI Key
UNLQTHFXAIISEP-UHFFFAOYSA-N
SMILES
[Cl].COC1=CC=C(NN)C=C1
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Appearance (Color)
White to pale pink to grey to purple
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text