Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads, Thermo Scientific Chemicals
Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
5000 g
Catalog number L06085.0B
also known as L06085-0B
Price (USD)/ Each
81.65
Online exclusive
91.00 
Save 9.35 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
81.65
Online exclusive
91.00 
Save 9.35 (10%)
Add to cart
Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads, Thermo Scientific Chemicals
Catalog numberL06085.0B
Price (USD)/ Each
81.65
Online exclusive
91.00 
Save 9.35 (10%)
-
Add to cart
Chemical Identifiers
CAS63231-69-6
IUPAC NameMolecular sieves 13X
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
CommentTapped bulk density 600 - 725 g/L
Particle SizeOversize: ≤5.0% @ >4.75mm (4 mesh)
CommentCO2 adsorption: ≥11.5% (45mbar, 25?C)
Particle SizeUndersize: ≤5.0% @ <2.36mm (8 mesh)
View more
Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only