trans-2-Pentenal, 96%, Thermo Scientific Chemicals
trans-2-Pentenal, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-Pentenal, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L06108.14
also known as L06108-14
Price (USD)/ Each
126.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
126.00
Add to cart
trans-2-Pentenal, 96%, Thermo Scientific Chemicals
Catalog numberL06108.14
Price (USD)/ Each
126.00
-
Add to cart
Chemical Identifiers
CAS1576-87-0
IUPAC Name(2E)-pent-2-enal
Molecular FormulaC5H8O
InChI KeyDTCCTIQRPGSLPT-ONEGZZNKSA-N
SMILESCC\C=C\C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4405-1.4465 @ 20°C
Appearance (Color)Clear colorless to white to yellow to red to black
Assay (GC)≥95.0%
Free acid (titration)≤3% (non-U.S. specification)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only