2,6-Dichloro-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals
2,6-Dichloro-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dichloro-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L06186.06
also known as L06186-06
Price (USD)/ Each
141.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
141.00
Add to cart
2,6-Dichloro-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals
Catalog numberL06186.06
Price (USD)/ Each
141.00
-
Add to cart
Chemical Identifiers
CAS5866-98-8
IUPAC Name2,6-dichloro-3-nitrobenzonitrile
Molecular FormulaC7H2Cl2N2O2
InChI KeyNSKVWZIEYFSHIM-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=C(Cl)C(C#N)=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)104.0-110.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only