Thermo Scientific Chemicals

4,5-Dichloroimidazole, 98%, Thermo Scientific Chemicals

Catalog number: L06262.14
25 g, Each
Thermo Scientific Chemicals

4,5-Dichloroimidazole, 98%, Thermo Scientific Chemicals

Catalog number: L06262.14
25 g, Each
Quantity
Catalog number: L06262.14
also known as L06262-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15965-30-7
Appearance (Color)
White to cream or yellow
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
176.0-183.0?C

Description

4,5-Dichloroimidazole has been used in the synthesis of: phosphinite complexes, 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dichloroimidazole has been used in the synthesis of: phosphinite complexes, 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text