Tris(trimethylsilyl) borate, 98%, Thermo Scientific Chemicals
Tris(trimethylsilyl) borate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(trimethylsilyl) borate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06277.06
also known as L06277-06
Price (USD)/ Each
30.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
30.90
Add to cart
Tris(trimethylsilyl) borate, 98%, Thermo Scientific Chemicals
Catalog numberL06277.06
Price (USD)/ Each
30.90
-
Add to cart
Chemical Identifiers
CAS4325-85-3
IUPAC Nametritrimethylsilyl borate
Molecular FormulaC9H27BO3Si3
InChI KeyYZYKZHPNRDIPFA-UHFFFAOYSA-N
SMILESC[Si](C)(C)OB(O[Si](C)(C)C)O[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.3840-1.3885 @ 20?C
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only