Thermo Scientific Chemicals

Glycolaldehyde diethyl acetal, stab. with ca 0.1% sodium carbonate, 98%, Thermo Scientific Chemicals

Catalog number: L06282.18
50 g, Each
Thermo Scientific Chemicals

Glycolaldehyde diethyl acetal, stab. with ca 0.1% sodium carbonate, 98%, Thermo Scientific Chemicals

Catalog number: L06282.18
50 g, Each
Quantity
Catalog number: L06282.18
also known as L06282-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
621-63-6
IUPAC Name
2,2-diethoxyethan-1-ol
Molecular Formula
C6H14O3
InChI Key
IKKUKDZKIIIKJK-UHFFFAOYSA-N
SMILES
CCOC(CO)OCC
Appearance (Color)
Clear, colorless
Assay (GC)
>97.5%
Refractive Index
1.4140-1.4180 @ 20?C
Form
Liquid

Description

Glycolaldehyde diethyl acetal is used in the preparation of neooxazolomycin, which is a part of the oxazolomycin family of antibiotics. It is also used in the preparation of pyrimidine based inhibitors of phosphodiesterase 7.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolaldehyde diethyl acetal is used in the preparation of neooxazolomycin, which is a part of the oxazolomycin family of antibiotics. It is also used in the preparation of pyrimidine based inhibitors of phosphodiesterase 7.

Solubility
Miscible with water, chloroform and ethyl acetate.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acid and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text