Thermo Scientific Chemicals

Ethyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog number: L06311.06
5 g, Each
Thermo Scientific Chemicals

Ethyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog number: L06311.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L06311.06
also known as L06311-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
541-41-3
IUPAC Name
ethyl carbonochloridate
Molecular Formula
C3H5ClO2
InChI Key
RIFGWPKJUGCATF-UHFFFAOYSA-N
SMILES
CCOC(Cl)=O
Form
Liquid
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.3930-1.3980 @ 20°C (non-U.S. specification)
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

Ethyl chloroformate is used as derivatization reagent to investigate stereochemical conversion of chiral non-steroidal anti-inflammatory drugs during derivatization reaction. It is also used with ammonia and cyanuric chloride to convert carboxylic acids to nitriles. It is used to develop flexible template strategy for the generation of nanometer-scale templates via dip-pen nanolithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl chloroformate is used as derivatization reagent to investigate stereochemical conversion of chiral non-steroidal anti-inflammatory drugs during derivatization reaction. It is also used with ammonia and cyanuric chloride to convert carboxylic acids to nitriles. It is used to develop flexible template strategy for the generation of nanometer-scale templates via dip-pen nanolithography.

Solubility
Not miscible in water.

Notes
Protect from heat. Store away from moisture and oxidizing agents. Store under dry inert gas. Refrigerate. Incompatible with heat, moisture and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text