Thermo Scientific Chemicals

L-Cysteine hydrochloride, anhydrous, 98%

Catalog number: L06328.14
25 g, Each
Thermo Scientific Chemicals

L-Cysteine hydrochloride, anhydrous, 98%

Catalog number: L06328.14
25 g, Each
Quantity
Catalog number: L06328.14
also known as L06328-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
52-89-1
IUPAC Name
(2R)-2-amino-3-sulfanylpropanoic acid hydrochloride
Molecular Formula
C3H8ClNO2S
InChI Key
IFQSXNOEEPCSLW-QYLAHFPANA-N
SMILES
Cl.N[C@@H](CS)C(O)=O
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
White
Comment
Specification differs for U.S. and non-U.S. material where indicated
Optical Rotation
6.1 ± 0.4° (C=5 in 5N HCl)
Form
Crystals or powder or crystalline powder

Description

L-Cysteine hydrochloride, anhydrous is widely used as additive in food production. It is used as an antioxidant to promote fermentation and keep the flavor. It is also used in cosmetics and animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• L-cysteine hydrochloride is a non-essential proteinogenic amino acid commonly used in chemical reactions, for instance, the thiol side chain in cysteine often participates in enzymatic reactions as a nucleophile.
Application
• L-cysteine hydrochloride plays an important role as a histidine ammonia-lyase inhibitor with the ability to interfere with the action of histidine ammonia-lyase.
• In chemical reactions, L-cysteine hydrochloride is used as a raw material for the production of S-carboxymethyl-L-cysteine, L-cysteine base, and N-acetyl-L-cysteine.
• This compound also has antioxidant activity.
• It has been used in laboratory protocols to separate dorsal root ganglion neurons.
• This compound is applied to study the inhibitory effect of L-cysteine hydrochloride hydrate in preventing enzymatic browning in apples.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text