3-Phenyl-2-propyn-1-ol, 97%, Thermo Scientific Chemicals
3-Phenyl-2-propyn-1-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Phenyl-2-propyn-1-ol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L06337.06
also known as L06337-06
Price (USD)/ Each
63.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
63.60
Add to cart
3-Phenyl-2-propyn-1-ol, 97%, Thermo Scientific Chemicals
Catalog numberL06337.06
Price (USD)/ Each
63.60
-
Add to cart
Chemical Identifiers
CAS1504-58-1
IUPAC Name3-phenylprop-2-yn-1-ol
Molecular FormulaC9H8O
InChI KeyNITUNGCLDSFVDL-UHFFFAOYSA-N
SMILESOCC#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 96.0%
Appearance (Color)Clear, colourless to yellow
Refractive Index1.5835 - 1.5885 @20?C
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only