Thermo Scientific Chemicals

2',6'-Dichloroacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: L06348.06
5 g, Each
Thermo Scientific Chemicals

2',6'-Dichloroacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: L06348.06
5 g, Each
Quantity
Catalog number: L06348.06
also known as L06348-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2040-05-3
IUPAC Name
1-(2,6-dichlorophenyl)ethan-1-one
Molecular Formula
C8H6Cl2O
InChI Key
HYBDSXBLGCQKRE-UHFFFAOYSA-N
SMILES
CC(=O)C1=C(Cl)C=CC=C1Cl
Melting Point (clear melt)
40.0-46.0?C
Appearance (Color)
White to cream to yellow
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid or flakes or granules

Description

2',6'-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,6′-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text