Thermo Scientific Chemicals

Trifluoroacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: L06374.AE
100 mL, Each
Thermo Scientific Chemicals

Trifluoroacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: L06374.AE
100 mL, Each
Quantity
Catalog number: L06374.AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-05-1
IUPAC Name
trifluoroacetic acid
Molecular Formula
C2HF3O2
InChI Key
DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)F
Comment
Material sourced in the U.S. and in other countries
Appearance (Color)
Clear colorless to pale yellow or pale pink
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. sourced material)
Identification (FTIR)
Conforms (non-U.S. sourced material)
Form
Fuming liquid

Description

The combination of properties like solubility in most of the solvents, volatility, catalytic property, and strong acidity with non-oxidizing nature makes it a widely used reagent in organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and performance products. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. TFA finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The combination of properties like solubility in most of the solvents, volatility, catalytic property, and strong acidity with non-oxidizing nature makes it a widely used reagent in organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and performance products. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. TFA finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

Solubility
Miscible with ether, acetone, ethanol, benzene, hexane and carbon tetrachloride.

Notes
Trifluoroacetic acid (TFA) is a stable, hygroscopic, volatile and low-boiling organic acid. Storage in a cool place is recommended.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text