Thermo Scientific Chemicals

Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals

Catalog number: L06407.06
5 g, Each
Thermo Scientific Chemicals

Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals

Catalog number: L06407.06
5 g, Each
Quantity
Catalog number: L06407.06
also known as L06407-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35192-74-6
IUPAC Name
sodium nonane-1-sulfonate
Molecular Formula
C9H19NaO3S
InChI Key
RUYRDULZOKULPK-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCS([O-])(=O)=O
Appearance (Color)
White
Form
Powder or crystalline powder
Assay from Suppliers CofA
≥97.5%: Ex-sulfated ash (U.S. specification)
Assay from Suppliers CofA
≥97.5%: Proton NMR (non-U.S.specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text