Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals
Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L06407.14
also known as L06407-14
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
Add to cart
Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals
Catalog numberL06407.14
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS35192-74-6
IUPAC Namesodium nonane-1-sulfonate
Molecular FormulaC9H19NaO3S
InChI KeyRUYRDULZOKULPK-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥97.5%: Ex-sulfated ash (U.S. specification)
Assay from Suppliers CofA≥97.5%: Proton NMR (non-U.S.specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Proton NMRConforms to structure (non-U.S. specification)
Water Content≤1.5% (U.S. specification)
View more
Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Tyagi, M. G.; Mathew, S. K. Lipoprotein associated phospholipase A2 enzyme; possible new roles and inhibition for therapeutic intervention. Int. J. Res. Med. Sci. 2014, 2 (3), 805-809.
  2. Pöllmann, H.; Stefan, S.; Stern, E. Synthesis, characterization and reaction behaviour of lamellar AFm phases with aliphatic sulfonate-anions. Cem. Concr. Res. 2006, 36 (11), 2039-2048.