Thermo Scientific Chemicals

4-Hydroxy-1-methylpiperidine, 98%, Thermo Scientific Chemicals

Catalog number: L06415.22
100 g, Each
Thermo Scientific Chemicals

4-Hydroxy-1-methylpiperidine, 98%, Thermo Scientific Chemicals

Catalog number: L06415.22
100 g, Each
Quantity
Catalog number: L06415.22
also known as L06415-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-52-5
IUPAC Name
1-methylpiperidin-4-ol
Molecular Formula
C6H13NO
InChI Key
BAUWRHPMUVYFOD-UHFFFAOYSA-N
SMILES
CN1CCC(O)CC1
Form
Liquid as melt
Refractive Index
1.4760-1.4810 as melt
Appearance (Color)
Clear colorless to brown
Assay (GC)
≥97.5%

Description

4-Hydroxy-1-methylpiperidine is used as reactant for optimization of Novobiocin scaffold to product antitumor agents, substitution about the rigidifying ring for histamine H4 receptor antagonist synthesis. It is also used as a reactant in the synthesis of CaMKII, VEGFR and FGFR kinase inhibitors and Phosphoinositide-3-kinase, Protein lysine methyltransferase G9a inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-1-methylpiperidine is used as reactant for optimization of Novobiocin scaffold to product antitumor agents, substitution about the rigidifying ring for histamine H4 receptor antagonist synthesis. It is also used as a reactant in the synthesis of CaMKII, VEGFR and FGFR kinase inhibitors and Phosphoinositide-3-kinase, Protein lysine methyltransferase G9a inhibitors.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under dry inert gas. Store away from moisture and oxidizing agents. Incompatible with water and oxidizing agents. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text