3',4',5'-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′,4′,5′-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Slightly soluble in water.
Notes
Incompatible materials are Strong oxidizing agents, Strong bases. Store away from strong oxidizing agents and stong bases. Avoid excessive heat and light exposure.
RUO – Research Use Only
Y Engelborghs; C Dumortier; A D’Hoore; A Vandecandelaere; T J Fitzgerald. Evidence for an alternative pathway for colchicine binding to tubulin, based on the binding kinetics of the constituent rings..Journal of Biological Chemistry.1993,268 (1), 107-112.