Thermo Scientific Chemicals

5-Chloro-m-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: L06485.06
5 g, Each
Thermo Scientific Chemicals

5-Chloro-m-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: L06485.06
5 g, Each
Quantity
Catalog number: L06485.06
also known as L06485-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33786-89-9
IUPAC Name
5-chlorobenzene-1,3-diamine
Molecular Formula
C6H7ClN2
InChI Key
VZNUCJOYPXKLTA-UHFFFAOYSA-N
SMILES
NC1=CC(Cl)=CC(N)=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
97.0-106.0?C
Appearance (Color)
Cream to yellow to orange to brown
Assay (GC)
≥96.0%

Description

5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text